Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFWD2
Entrez Gene ID
64326
Gene Name
ring finger and WD repeat domain 2
Gene Aliases
COP1, RNF200
Chromosome Location
Chr.1: 175944826 - 176207279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001740.3 NP_001001740.1 10 - 11 1476 103
NM_001286644.1 NP_001273573.1 9 - 10 1351 103
NM_022457.6 NP_071902.2 11 - 12 1548 103
XM_005245447.2 XP_005245504.1 11 - 12 1571 103
XM_005245448.2 XP_005245505.1 10 - 11 1523 103
XM_006711487.2 XP_006711550.1 10 - 11 1514 103
XM_011509881.1 XP_011508183.1 12 - 13 1637 103
XM_017002059.1 XP_016857548.1 12 - 13 1637 103
XM_017002060.1 XP_016857549.1 12 - 13 1625 103
XM_017002061.1 XP_016857550.1 11 - 12 1583 103
XM_017002062.1 XP_016857551.1 11 - 12 1577 103
XM_017002063.1 XP_016857552.1 11 - 12 1571 103
XM_017002064.1 XP_016857553.1 11 - 12 1568 103
XM_017002065.1 XP_016857554.1 10 - 11 1523 103
XM_017002066.1 XP_016857555.1 10 - 11 1514 103
XM_017002067.1 XP_016857556.1 10 - 11 1511 103
XM_017002068.1 XP_016857557.1 10 - 11 1508 103
XM_017002069.1 XP_016857558.1 6 - 7 1151 103
XM_017002070.1 XP_016857559.1 11 - 12 1343 103
XM_017002071.1 XP_016857560.1 9 - 10 732 103
XM_017002072.1 XP_016857561.1 9 - 10 730 103
XM_017002073.1 XP_016857562.1 8 - 9 672 103
XM_017002074.1 XP_016857563.1 8 - 9 670 103
XM_017002075.1 XP_016857564.1 8 - 9 752 103
XM_017002076.1 XP_016857565.1 9 - 10 735 103
XM_017002077.1 XP_016857566.1 7 - 8 527 103
XM_017002078.1 XP_016857567.1 9 - 10 800 103
XM_017002079.1 XP_016857568.1 9 - 10 1215 103
XM_017002080.1 XP_016857569.1 5 - 6 454 103
GenBank mRNA AF508940.1 - 11 - 12 1282 103
AF527539.1 - 11 - 12 1545 103
AK001278.1 - 9 - 10 787 103
AK314750.1 - 11 - 12 1480 103
AY509921.1 - 10 - 11 1210 103
BC039723.1 - 9 - 10 798 103
BC094728.1 - 11 - 12 1536 103
BX537932.1 - 1 - 2 110 103
KJ534928.1 - 11 - 12 1282 103
KJ534929.1 - 8 - 9 1087 103
KJ534930.1 - 10 - 11 1222 103
KJ534931.1 - 10 - 11 1213 103
KJ534932.1 - 11 - 12 1270 103
KJ534933.1 - 5 - 6 781 103
KJ535061.1 - 8 - 9 987 103
KJ535076.1 - 10 - 11 1184 103

View Full Product Details