The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052867.2 | NP_443099.1 | 19 - 20 | 2482 | 62 |
XM_011521067.2 | XP_011519369.1 | 19 - 20 | 2422 | 62 | |
XM_011521068.2 | XP_011519370.1 | 19 - 20 | 2405 | 62 | |
XM_011521069.2 | XP_011519371.1 | 18 - 19 | 2335 | 62 | |
XM_017020534.1 | XP_016876023.1 | 18 - 19 | 2366 | 62 | |
XM_017020535.1 | XP_016876024.1 | 18 - 19 | 2308 | 62 | |
XM_017020536.1 | XP_016876025.1 | 14 - 15 | 2234 | 62 | |
XM_017020537.1 | XP_016876026.1 | 13 - 14 | 1623 | 62 | |
GenBank mRNA | AY141972.1 | - | 19 - 20 | 2376 | 62 |