The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052867.2 | NP_443099.1 | 30 - 31 | 3675 | 103 |
XM_011521067.2 | XP_011519369.1 | 30 - 31 | 3615 | 103 | |
XM_011521068.2 | XP_011519370.1 | 30 - 31 | 3598 | 103 | |
XM_011521069.2 | XP_011519371.1 | 29 - 30 | 3528 | 103 | |
XM_017020534.1 | XP_016876023.1 | 29 - 30 | 3559 | 103 | |
XM_017020535.1 | XP_016876024.1 | 29 - 30 | 3501 | 103 | |
XM_017020536.1 | XP_016876025.1 | 25 - 26 | 3427 | 103 | |
XM_017020537.1 | XP_016876026.1 | 24 - 25 | 2816 | 103 | |
GenBank mRNA | AK126621.1 | - | 11 - 12 | 1139 | 103 |
AY141972.1 | - | 30 - 31 | 3569 | 103 | |
DA128114.1 | - | 1 - 2 | 107 | 103 |