The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001949.4 | NP_001940.1 | 1 - 2 | 726 | 146 |
XM_011514324.2 | XP_011512626.1 | 1 - 2 | 1096 | 146 | |
GenBank mRNA | Y10479.1 | - | 1 - 2 | 465 | 146 |