The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004533.3 | NP_004524.3 | 2 - 3 | 163 | 107 |
GenBank mRNA | BC130536.1 | - | 2 - 3 | 130 | 107 |
BC136389.1 | - | 2 - 3 | 130 | 107 | |
CF553029.1 | - | 2 - 3 | 163 | 107 | |
X73113.1 | - | 2 - 3 | 131 | 107 |