The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005228.3 | NP_005219.2 | 18 - 19 | 2426 | 126 |
GenBank mRNA | AB209442.1 | - | 18 - 19 | 2325 | 126 |
AK290352.1 | - | 18 - 19 | 2226 | 126 | |
AK294750.1 | - | 18 - 19 | 2204 | 126 | |
AY550966.1 | - | 18 - 19 | 2226 | 126 | |
BC094761.1 | - | 17 - 18 | 2302 | 126 | |
DQ088980.1 | - | 18 - 19 | 2226 | 126 | |
GU255993.1 | - | 18 - 19 | 2223 | 126 | |
HQ912715.1 | - | 17 - 18 | 2291 | 126 | |
X00588.1 | - | 18 - 19 | 2366 | 126 | |
X00663.1 | - | 15 - 16 | 1754 | 126 |