The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128226.2 | NP_001121698.1 | 13 - 14 | 132 | |
NM_001322348.1 | NP_001309277.1 | 12 - 13 | 132 | ||
NM_001322349.1 | NP_001309278.1 | 14 - 15 | 132 | ||
NM_014953.4 | NP_055768.3 | 13 - 14 | 132 | ||
GenBank mRNA | AB001743.1 | - | 13 - 14 | 1782 | 132 |
AB023225.1 | - | 13 - 14 | 1758 | 132 | |
AF330044.1 | - | 13 - 14 | 1791 | 132 | |
AK025634.1 | - | 11 - 12 | 1361 | 132 | |
AK314715.1 | - | 13 - 14 | 1867 | 132 | |
AL080158.1 | - | 7 - 8 | 132 | ||
AL832266.1 | - | 13 - 14 | 1844 | 132 | |
BC038101.1 | - | 14 - 15 | 2090 | 132 | |
BC056143.1 | - | 13 - 14 | 1856 | 132 |