The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000256.3 | NP_000247.2 | 16 - 17 | 1513 | 136 |
GenBank mRNA | AK297843.1 | - | 16 - 17 | 1513 | 136 |
AL691822.1 | - | 5 - 6 | 372 | 136 | |
BC136543.1 | - | 16 - 17 | 1490 | 136 | |
BC136546.1 | - | 16 - 17 | 1490 | 136 | |
BC142685.1 | - | 16 - 17 | 1493 | 136 | |
BC151211.1 | - | 16 - 17 | 1496 | 136 | |
EF560722.1 | - | 16 - 17 | 1514 | 136 | |
EU747718.1 | - | 16 - 17 | 1513 | 136 | |
EU747719.1 | - | 16 - 17 | 1513 | 136 |