The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012454.3 | NP_036586.2 | 14 - 15 | 3375 | 96 |
GenBank mRNA | AF120323.1 | - | 10 - 11 | 1345 | 96 |
AK123859.1 | - | 15 - 16 | 3470 | 96 | |
AK302860.1 | - | 10 - 11 | 1286 | 96 | |
AL122086.1 | - | 9 - 10 | 1204 | 96 | |
BC137275.1 | - | 13 - 14 | 3248 | 96 |