The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012454.3 | NP_036586.2 | 3 - 4 | 1399 | 61 |
GenBank mRNA | AK123859.1 | - | 4 - 5 | 1494 | 61 |
BC137275.1 | - | 2 - 3 | 1272 | 61 |