The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082618.1 | NP_001076087.1 | 3 - 4 | 404 | 102 |
NM_001177608.1 | NP_001171079.1 | 4 - 5 | 785 | 102 | |
NM_001772.3 | NP_001763.3 | 4 - 5 | 785 | 102 | |
XM_011527531.2 | XP_011525833.1 | 10 - 11 | 1859 | 102 | |
XM_011527532.2 | XP_011525834.1 | 9 - 10 | 1822 | 102 | |
XM_017027508.1 | XP_016882997.1 | 10 - 11 | 1995 | 102 | |
XM_017027510.1 | XP_016882999.1 | 8 - 9 | 1566 | 102 | |
GenBank mRNA | AK293934.1 | - | 3 - 4 | 419 | 102 |
AK304810.1 | - | 4 - 5 | 749 | 102 | |
AK309558.1 | - | 4 - 5 | 766 | 102 | |
AY162464.1 | - | 3 - 4 | 372 | 102 | |
BC028152.1 | - | 4 - 5 | 750 | 102 | |
KX139463.1 | - | 3 - 4 | 364 | 102 | |
M23197.1 | - | 4 - 5 | 757 | 102 |