Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFL2
Entrez Gene ID
1073
Gene Name
cofilin 2
Gene Aliases
NEM7
Chromosome Location
Chr.14: 34710382 - 34714823 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011536363.2 XP_011534665.1 1 - 2 87 156

View Full Product Details