Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLASP1
Entrez Gene ID
23332
Gene Name
cytoplasmic linker associated protein 1
Gene Aliases
MAST1
Chromosome Location
Chr.2: 121337776 - 121649476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142273.1 NP_001135745.1 15 - 16 1835 146
NM_001142274.1 NP_001135746.1 15 - 16 1835 146
NM_001207051.1 NP_001193980.1 15 - 16 1835 146
NM_015282.2 NP_056097.1 15 - 16 1835 146
XM_006712377.1 XP_006712440.1 15 - 16 1806 146
XM_006712381.1 XP_006712444.1 15 - 16 1806 146
XM_006712382.1 XP_006712445.1 15 - 16 1806 146
XM_006712383.1 XP_006712446.1 15 - 16 1806 146
XM_006712385.1 XP_006712448.1 9 - 10 878 146
XM_011510848.1 XP_011509150.1 15 - 16 1806 146
XM_011510849.1 XP_011509151.1 15 - 16 1806 146
XM_017003657.1 XP_016859146.1 15 - 16 1806 146
XM_017003658.1 XP_016859147.1 15 - 16 1806 146
XM_017003659.1 XP_016859148.1 15 - 16 1806 146
XM_017003660.1 XP_016859149.1 15 - 16 1806 146
XM_017003661.1 XP_016859150.1 15 - 16 1806 146
XM_017003662.1 XP_016859151.1 15 - 16 1806 146
XM_017003663.1 XP_016859152.1 15 - 16 1806 146
XM_017003664.1 XP_016859153.1 15 - 16 1806 146
XM_017003665.1 XP_016859154.1 15 - 16 1806 146
XM_017003666.1 XP_016859155.1 15 - 16 1806 146
XM_017003667.1 XP_016859156.1 15 - 16 1806 146
XM_017003668.1 XP_016859157.1 15 - 16 1806 146
XM_017003669.1 XP_016859158.1 15 - 16 1806 146
XM_017003670.1 XP_016859159.1 15 - 16 1806 146
XM_017003671.1 XP_016859160.1 15 - 16 1806 146
XM_017003672.1 XP_016859161.1 15 - 16 1806 146
XM_017003673.1 XP_016859162.1 15 - 16 1806 146
XM_017003674.1 XP_016859163.1 15 - 16 1806 146
XM_017003675.1 XP_016859164.1 15 - 16 1806 146
XM_017003676.1 XP_016859165.1 8 - 9 2640 146
XM_017003677.1 XP_016859166.1 8 - 9 2640 146
XM_017003678.1 XP_016859167.1 8 - 9 2639 146
XM_017003679.1 XP_016859168.1 8 - 9 2640 146
XM_017003680.1 XP_016859169.1 8 - 9 2640 146
XM_017003681.1 XP_016859170.1 8 - 9 2639 146
XM_017003682.1 XP_016859171.1 8 - 9 2639 146
XM_017003683.1 XP_016859172.1 8 - 9 2638 146
XM_017003684.1 XP_016859173.1 8 - 9 1621 146
XM_017003685.1 XP_016859174.1 8 - 9 2638 146
XM_017003686.1 XP_016859175.1 8 - 9 2638 146
XM_017003687.1 XP_016859176.1 8 - 9 1621 146
XM_017003688.1 XP_016859177.1 8 - 9 2638 146
XM_017003689.1 XP_016859178.1 8 - 9 1620 146
XM_017003690.1 XP_016859179.1 15 - 16 1806 146
XM_017003691.1 XP_016859180.1 15 - 16 1806 146
GenBank mRNA AB014522.1 - 7 - 8 698 146
AF347693.1 - 14 - 15 1610 146
AK074338.1 - 2 - 3 118 146
AK091140.1 - 15 - 16 1796 146
AK297259.1 - 8 - 9 789 146
AK299331.1 - 10 - 11 1357 146
AK302393.1 - 9 - 10 895 146
BC112940.1 - 15 - 16 1779 146
BC132723.1 - 15 - 16 1797 146
BC144107.1 - 15 - 16 1797 146
CR933722.1 - 8 - 9 787 146

View Full Product Details