Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CLASP1
- Entrez Gene ID
-
23332
- Gene Name
- cytoplasmic linker associated protein 1
- Gene Aliases
-
MAST1
- Chromosome Location
-
Chr.2: 121337776 - 121649476 on Build GRCh38
View Full Product Details