The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127198.2 | NP_001120670.1 | 1 - 2 | 180 | 125 |
XM_017024107.1 | XP_016879596.1 | 1 - 2 | 158 | 125 | |
GenBank mRNA | AK074065.1 | - | 1 - 2 | 83 | 125 |
AK096076.1 | - | 1 - 2 | 116 | 125 | |
AK303584.1 | - | 1 - 2 | 141 | 125 | |
HY100488.1 | - | 1 - 2 | 180 | 125 |