The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130969.1 | NP_001124441.1 | 8 - 9 | 1154 | 97 |
NM_001130970.1 | NP_001124442.1 | 7 - 8 | 1085 | 97 | |
NM_001130971.1 | NP_001124443.1 | 6 - 7 | 1079 | 97 | |
NM_015537.4 | NP_056352.3 | 7 - 8 | 1148 | 97 | |
XM_011518497.2 | XP_011516799.1 | 3 - 4 | 268 | 97 | |
GenBank mRNA | AK027474.1 | - | 5 - 6 | 419 | 97 |
AK074602.1 | - | 7 - 8 | 1116 | 97 | |
AK292030.1 | - | 3 - 4 | 220 | 97 | |
AK303703.1 | - | 5 - 6 | 276 | 97 | |
AK303976.1 | - | 3 - 4 | 315 | 97 | |
AK316046.1 | - | 5 - 6 | 470 | 97 | |
AL389944.1 | - | 3 - 4 | 202 | 97 | |
AY255128.1 | - | 8 - 9 | 1154 | 97 | |
AY255129.1 | - | 7 - 8 | 1148 | 97 | |
AY255130.1 | - | 7 - 8 | 1085 | 97 | |
AY255132.1 | - | 8 - 9 | 1154 | 97 | |
BC016862.2 | - | 3 - 4 | 195 | 97 | |
BC072412.1 | - | 7 - 8 | 1133 | 97 | |
BC110498.2 | - | 6 - 7 | 946 | 97 |