Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCN9A
Entrez Gene ID
6335
Gene Name
sodium voltage-gated channel alpha subunit 9
Gene Aliases
ETHA, FEB3B, GEFSP7, HSAN2D, NE-NA, NENA, Nav1.7, PN1, SFNP
Chromosome Location
Chr.2: 166195185 - 166375987 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002977.3 NP_002968.1 8 - 9 1308 75
XM_005246757.2 XP_005246814.1 8 - 9 1281 75
XM_011511616.2 XP_011509918.1 8 - 9 1242 75
XM_011511617.2 XP_011509919.1 8 - 9 1281 75
XM_011511618.2 XP_011509920.1 8 - 9 1281 75
XM_011511619.2 XP_011509921.1 8 - 9 1281 75
XM_017004668.1 XP_016860157.1 7 - 8 810 75
XM_017004669.1 XP_016860158.1 3 - 4 368 75
GenBank mRNA AK307549.1 - 8 - 9 1281 75
AY682084.1 - 5 - 6 562 75
AY682085.1 - 5 - 6 562 75
AY682086.1 - 5 - 6 562 75
DQ857292.1 - 7 - 8 1008 75
X82835.1 - 7 - 8 1015 75

View Full Product Details