The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000088.3 | NP_000079.2 | 12 - 13 | 987 | 77 |
XM_005257058.3 | XP_005257115.2 | 12 - 13 | 996 | 77 | |
XM_005257059.4 | XP_005257116.2 | 12 - 13 | 996 | 77 | |
XM_011524341.1 | XP_011522643.1 | 12 - 13 | 996 | 77 | |
GenBank mRNA | AB209597.1 | - | 12 - 13 | 962 | 77 |
BC036531.2 | - | 12 - 13 | 977 | 77 | |
JQ236861.1 | - | 12 - 13 | 861 | 77 | |
M36546.1 | - | 12 - 13 | 944 | 77 | |
X07884.1 | - | 12 - 13 | 944 | 77 | |
Z74615.1 | - | 12 - 13 | 980 | 77 |