The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000088.3 | NP_000079.2 | 50 - 51 | 4372 | 161 |
XM_005257058.3 | XP_005257115.2 | 48 - 49 | 4111 | 161 | |
XM_005257059.4 | XP_005257116.2 | 37 - 38 | 3463 | 161 | |
XM_011524341.1 | XP_011522643.1 | 47 - 48 | 4183 | 161 | |
GenBank mRNA | AB209597.1 | - | 50 - 51 | 4347 | 161 |
BC036531.2 | - | 50 - 51 | 4362 | 161 | |
JQ236861.1 | - | 50 - 51 | 4238 | 161 | |
K01228.1 | - | 32 - 33 | 2974 | 161 | |
S64596.1 | - | 3 - 4 | 706 | 161 | |
Z74615.1 | - | 50 - 51 | 4365 | 161 |