Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REV3L
Entrez Gene ID
5980
Gene Name
REV3 like, DNA directed polymerase zeta catalytic subunit
Gene Aliases
POLZ, REV3
Chromosome Location
Chr.6: 111299031 - 111483715 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286431.1 NP_001273360.1 5 - 6 1022 146
NM_001286432.1 NP_001273361.1 4 - 5 915 146
NM_002912.4 NP_002903.3 3 - 4 787 146
XM_011536028.1 XP_011534330.1 2 - 3 878 146
XM_011536029.2 XP_011534331.1 2 - 3 878 146
XM_011536030.2 XP_011534332.1 2 - 3 878 146
XM_011536031.2 XP_011534333.1 3 - 4 1006 146
XM_011536032.2 XP_011534334.1 3 - 4 5596 146
XM_011536036.2 XP_011534338.1 2 - 3 878 146
XM_017011152.1 XP_016866641.1 3 - 4 1006 146
XM_017011153.1 XP_016866642.1 3 - 4 549 146
XM_017011154.1 XP_016866643.1 3 - 4 434 146
XM_017011155.1 XP_016866644.1 2 - 3 878 146
GenBank mRNA AF035537.1 - 1 - 2 124 146
AF058701.1 - 3 - 4 414 146
AF071798.1 - 2 - 3 652 146
AF078695.1 - 4 - 5 917 146
AF157476.1 - 2 - 3 360 146
AF179428.1 - 3 - 4 488 146
AF179429.1 - 4 - 5 595 146
AK310949.1 - 4 - 5 911 146
BC035823.1 - 2 - 3 586 146

View Full Product Details