The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321038.1 | NP_001307967.1 | 6 - 7 | 945 | 141 |
NM_001321039.1 | NP_001307968.1 | 6 - 7 | 1177 | 141 | |
NM_013296.4 | NP_037428.3 | 6 - 7 | 1177 | 141 | |
XM_006710589.2 | XP_006710652.1 | 5 - 6 | 1636 | 141 | |
XM_011541301.2 | XP_011539603.1 | 7 - 8 | 1454 | 141 | |
XM_011541302.2 | XP_011539604.1 | 7 - 8 | 2116 | 141 | |
XM_011541303.2 | XP_011539605.1 | 6 - 7 | 1872 | 141 | |
XM_017001097.1 | XP_016856586.1 | 7 - 8 | 2022 | 141 | |
XM_017001098.1 | XP_016856587.1 | 7 - 8 | 1790 | 141 | |
GenBank mRNA | AB445462.1 | - | 6 - 7 | 1177 | 141 |
AK295563.1 | - | 6 - 7 | 764 | 141 | |
AK314249.1 | - | 6 - 7 | 1173 | 141 | |
AY136740.1 | - | 5 - 6 | 660 | 141 | |
BC027732.2 | - | 6 - 7 | 953 | 141 | |
CR456786.1 | - | 5 - 6 | 660 | 141 | |
KX085485.1 | - | 6 - 7 | 1177 | 141 | |
U54999.1 | - | 5 - 6 | 833 | 141 |