Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP5F1
Entrez Gene ID
515
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1
Gene Aliases
PIG47
Chromosome Location
Chr.1: 111449121 - 111461918 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK308266.1 - 1 - 2 116 142

View Full Product Details