The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271641.1 | NP_001258570.1 | 9 - 10 | 1337 | 78 |
NM_014341.2 | NP_055156.1 | 9 - 10 | 1286 | 78 | |
NR_130739.1 | 9 - 10 | 1379 | 78 | ||
XM_005248978.1 | XP_005249035.1 | 9 - 10 | 1337 | 78 | |
XM_005248979.1 | XP_005249036.1 | 9 - 10 | 1286 | 78 | |
XM_011514450.1 | XP_011512752.1 | 8 - 9 | 794 | 78 | |
XM_017010642.1 | XP_016866131.1 | 8 - 9 | 845 | 78 | |
XM_017010643.1 | XP_016866132.1 | 8 - 9 | 840 | 78 | |
GenBank mRNA | AF176006.3 | - | 9 - 10 | 908 | 78 |
AF189289.1 | - | 9 - 10 | 918 | 78 | |
AF192559.3 | - | 9 - 10 | 959 | 78 | |
AK022783.1 | - | 9 - 10 | 906 | 78 | |
AK130842.1 | - | 9 - 10 | 990 | 78 | |
AK225628.1 | - | 9 - 10 | 890 | 78 | |
AK303318.1 | - | 8 - 9 | 707 | 78 | |
AK315067.1 | - | 9 - 10 | 1287 | 78 | |
AY358519.1 | - | 6 - 7 | 367 | 78 | |
AY947344.1 | - | 9 - 10 | 877 | 78 | |
BC000702.2 | - | 9 - 10 | 838 | 78 | |
BC006507.2 | - | 9 - 10 | 638 | 78 | |
BC009962.2 | - | 9 - 10 | 887 | 78 | |
BC040659.1 | - | 9 - 10 | 905 | 78 | |
BC110914.1 | - | 9 - 10 | 929 | 78 | |
BC125056.1 | - | 9 - 10 | 903 | 78 | |
BC153873.1 | - | 9 - 10 | 959 | 78 |