Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD83
Entrez Gene ID
9308
Gene Name
CD83 molecule
Gene Aliases
BL11, HB15
Chromosome Location
Chr.6: 14117256 - 14136918 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040280.1 NP_001035370.1 1 - 2 210 78
NM_004233.3 NP_004224.1 1 - 2 210 78
GenBank mRNA AK223315.1 - 1 - 2 72 78
AK290426.1 - 1 - 2 72 78
AK309541.1 - 1 - 2 70 78
BC030830.1 - 1 - 2 164 78
BT019498.1 - 1 - 2 32 78
CR457019.1 - 1 - 2 32 78
DB178710.1 - 1 - 2 210 78
Z11697.1 - 1 - 2 42 78

View Full Product Details