The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145038.4 | NP_659475.2 | 16 - 17 | 2246 | 71 |
XM_005264637.3 | XP_005264694.1 | 15 - 16 | 2054 | 71 | |
XM_005264638.3 | XP_005264695.1 | 12 - 13 | 1601 | 71 | |
XM_017005271.1 | XP_016860760.1 | 15 - 16 | 2014 | 71 | |
GenBank mRNA | AK057222.1 | - | 16 - 17 | 2242 | 71 |
AK289953.1 | - | 16 - 17 | 2229 | 71 | |
AL833892.1 | - | 15 - 16 | 1980 | 71 | |
BC009488.2 | - | 15 - 16 | 1969 | 71 |