The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145038.4 | NP_659475.2 | 1 - 2 | 236 | 71 |
XM_005264637.3 | XP_005264694.1 | 1 - 2 | 228 | 71 | |
XM_017005271.1 | XP_016860760.1 | 1 - 2 | 144 | 71 | |
GenBank mRNA | AK057222.1 | - | 1 - 2 | 232 | 71 |
AK289953.1 | - | 1 - 2 | 219 | 71 | |
AK307094.1 | - | 1 - 2 | 210 | 71 | |
DB021259.1 | - | 1 - 2 | 236 | 71 |