The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322340.1 | NP_001309269.1 | 11 - 12 | 1437 | 119 |
NM_001322341.1 | NP_001309270.1 | 12 - 13 | 1541 | 119 | |
NM_001322342.1 | NP_001309271.1 | 11 - 12 | 941 | 119 | |
NM_001322343.1 | NP_001309272.1 | 11 - 12 | 1000 | 119 | |
NM_022779.8 | NP_073616.6 | 11 - 12 | 1437 | 119 | |
NM_138620.1 | NP_619526.1 | 11 - 12 | 1437 | 119 | |
NR_136309.1 | 11 - 12 | 1437 | 119 | ||
XM_005272207.4 | XP_005272264.1 | 11 - 12 | 1124 | 119 | |
XM_006717236.3 | XP_006717299.1 | 11 - 12 | 1439 | 119 | |
XM_011518921.2 | XP_011517223.1 | 11 - 12 | 1439 | 119 | |
XM_011518922.2 | XP_011517224.1 | 11 - 12 | 1439 | 119 | |
GenBank mRNA | AF335568.1 | - | 11 - 12 | 1438 | 119 |
AF335569.1 | - | 11 - 12 | 1438 | 119 | |
AF427339.1 | - | 11 - 12 | 1438 | 119 | |
AK023695.1 | - | 7 - 8 | 510 | 119 | |
AK027002.1 | - | 11 - 12 | 1021 | 119 | |
AK027484.1 | - | 11 - 12 | 1438 | 119 | |
AK225121.1 | - | 11 - 12 | 992 | 119 | |
AK225350.1 | - | 11 - 12 | 1022 | 119 | |
AK302670.1 | - | 11 - 12 | 1174 | 119 | |
BC012726.2 | - | 11 - 12 | 1063 | 119 |