The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322341.1 | NP_001309270.1 | 16 - 17 | 2063 | 65 |
NM_001322342.1 | NP_001309271.1 | 15 - 16 | 1463 | 65 | |
NM_001322343.1 | NP_001309272.1 | 15 - 16 | 1522 | 65 | |
NM_022779.8 | NP_073616.6 | 15 - 16 | 1959 | 65 | |
XM_005272207.4 | XP_005272264.1 | 15 - 16 | 1646 | 65 | |
XM_006717236.3 | XP_006717299.1 | 15 - 16 | 1961 | 65 | |
XM_011518921.2 | XP_011517223.1 | 15 - 16 | 1961 | 65 | |
XM_011518922.2 | XP_011517224.1 | 15 - 16 | 1961 | 65 | |
GenBank mRNA | AF335568.1 | - | 15 - 16 | 1960 | 65 |
AF427339.1 | - | 15 - 16 | 1960 | 65 | |
AK023695.1 | - | 11 - 12 | 1032 | 65 | |
AK027002.1 | - | 15 - 16 | 1543 | 65 | |
AK225121.1 | - | 15 - 16 | 1514 | 65 | |
AK225350.1 | - | 15 - 16 | 1544 | 65 | |
AK302670.1 | - | 15 - 16 | 1696 | 65 | |
BC012726.2 | - | 15 - 16 | 1585 | 65 |