The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198978.1 | NP_001185907.1 | 4 - 5 | 369 | 66 |
NM_001198979.1 | NP_001185908.1 | 4 - 5 | 453 | 66 | |
NM_001198980.1 | NP_001185909.1 | 4 - 5 | 384 | 66 | |
NM_022733.2 | NP_073570.1 | 4 - 5 | 811 | 66 | |
XM_011541960.1 | XP_011540262.1 | 5 - 6 | 818 | 66 | |
GenBank mRNA | AK298702.1 | - | 4 - 5 | 349 | 66 |
AK303975.1 | - | 4 - 5 | 357 | 66 | |
AK308280.1 | - | 4 - 5 | 453 | 66 | |
AK313105.1 | - | 4 - 5 | 758 | 66 | |
AK316218.1 | - | 4 - 5 | 384 | 66 | |
AL133206.1 | - | 4 - 5 | 325 | 66 | |
AL137764.1 | - | 4 - 5 | 338 | 66 | |
BC014512.1 | - | 2 - 3 | 150 | 66 | |
BC021133.1 | - | 4 - 5 | 461 | 66 | |
DC411665.1 | - | 4 - 5 | 369 | 66 |