The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135955.1 | NP_001129427.1 | 11 - 12 | 2595 | 112 |
NM_001135956.1 | NP_001129428.1 | 10 - 11 | 2400 | 112 | |
GenBank mRNA | AF063823.1 | - | 11 - 12 | 2595 | 112 |
AF063825.1 | - | 10 - 11 | 2400 | 112 | |
AF421359.1 | - | 10 - 11 | 2360 | 112 |