The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318765.1 | NP_001305694.1 | 6 - 7 | 870 | 144 |
NM_032124.4 | NP_115500.1 | 6 - 7 | 813 | 144 | |
XM_006722557.2 | XP_006722620.1 | 7 - 8 | 905 | 144 | |
XM_011526227.2 | XP_011524529.1 | 7 - 8 | 889 | 144 | |
XM_011526228.2 | XP_011524530.1 | 7 - 8 | 887 | 144 | |
XM_017026039.1 | XP_016881528.1 | 7 - 8 | 926 | 144 | |
GenBank mRNA | AK054590.1 | - | 6 - 7 | 850 | 144 |
AK292098.1 | - | 6 - 7 | 813 | 144 | |
AL136681.1 | - | 6 - 7 | 804 | 144 | |
BC011894.2 | - | 6 - 7 | 834 | 144 | |
BC033031.1 | - | 6 - 7 | 849 | 144 | |
BC038100.2 | - | 6 - 7 | 813 | 144 | |
CR533487.1 | - | 5 - 6 | 680 | 144 | |
FJ224322.1 | - | 6 - 7 | 804 | 144 |