The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005720.3 | NP_005711.1 | 5 - 6 | 618 | 61 |
XM_006715825.1 | XP_006715888.1 | 5 - 6 | 629 | 61 | |
XM_006715826.1 | XP_006715889.1 | 5 - 6 | 613 | 61 | |
GenBank mRNA | AF006084.1 | - | 5 - 6 | 578 | 61 |
AK314818.1 | - | 5 - 6 | 563 | 61 | |
AU143525.1 | - | 5 - 6 | 618 | 61 | |
AY007111.1 | - | 3 - 4 | 448 | 61 | |
BC002562.2 | - | 5 - 6 | 551 | 61 | |
BC002988.1 | - | 5 - 6 | 587 | 61 | |
BC007555.1 | - | 5 - 6 | 549 | 61 |