Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGAP4
Entrez Gene ID
393
Gene Name
Rho GTPase activating protein 4
Gene Aliases
C1, RGC1, RhoGAP4, SrGAP4, p115
Chromosome Location
Chr.X: 153907376 - 153926260 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164741.1 NP_001158213.1 19 - 20 2406 82
NM_001666.4 NP_001657.3 18 - 19 2286 82
GenBank mRNA AY183123.1 - 15 - 16 1852 82
BC023626.2 - 4 - 5 460 82
BC052303.1 - 19 - 20 2406 82
D50921.1 - 18 - 19 2217 82
X78817.1 - 18 - 19 2270 82

View Full Product Details