The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164741.1 | NP_001158213.1 | 3 - 4 | 498 | 92 |
NM_001666.4 | NP_001657.3 | 3 - 4 | 498 | 92 | |
GenBank mRNA | AK294562.1 | - | 3 - 4 | 423 | 92 |
AY183123.1 | - | 3 - 4 | 482 | 92 | |
BC052303.1 | - | 3 - 4 | 498 | 92 | |
D50921.1 | - | 3 - 4 | 429 | 92 | |
X78817.1 | - | 3 - 4 | 482 | 92 |