Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDC37L1
Entrez Gene ID
55664
Gene Name
cell division cycle 37 like 1
Gene Aliases
CDC37B, HARC
Chromosome Location
Chr.9: 4679553 - 4708399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017913.3 NP_060383.2 4 - 5 839 142
GenBank mRNA AK000497.1 - 4 - 5 834 142
AK000646.1 - 4 - 5 723 142
AK225510.1 - 4 - 5 723 142
BC014133.2 - 4 - 5 659 142

View Full Product Details