The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004586.2 | NP_004577.1 | 16 - 17 | 1447 | 151 |
XM_005274573.2 | XP_005274630.1 | 16 - 17 | 1448 | 151 | |
XM_005274577.2 | XP_005274634.1 | 15 - 16 | 1384 | 151 | |
XM_006724507.3 | XP_006724570.1 | 18 - 19 | 1550 | 151 | |
XM_011545555.1 | XP_011543857.1 | 16 - 17 | 1469 | 151 | |
XM_011545556.1 | XP_011543858.1 | 16 - 17 | 1466 | 151 | |
XM_011545557.2 | XP_011543859.1 | 18 - 19 | 1587 | 151 | |
XM_011545558.2 | XP_011543860.1 | 18 - 19 | 1651 | 151 | |
XM_011545559.1 | XP_011543861.1 | 18 - 19 | 1648 | 151 | |
XM_011545560.1 | XP_011543862.1 | 17 - 18 | 1503 | 151 | |
XM_011545561.1 | XP_011543863.1 | 16 - 17 | 1429 | 151 | |
XM_011545562.1 | XP_011543864.1 | 15 - 16 | 1405 | 151 | |
XM_011545563.2 | XP_011543865.1 | 18 - 19 | 1567 | 151 | |
XM_017029713.1 | XP_016885202.1 | 18 - 19 | 1555 | 151 | |
XM_017029714.1 | XP_016885203.1 | 18 - 19 | 1631 | 151 | |
XM_017029715.1 | XP_016885204.1 | 18 - 19 | 1630 | 151 | |
XM_017029716.1 | XP_016885205.1 | 18 - 19 | 1705 | 151 | |
XM_017029717.1 | XP_016885206.1 | 16 - 17 | 1411 | 151 | |
XM_017029718.1 | XP_016885207.1 | 15 - 16 | 1387 | 151 | |
XM_017029719.1 | XP_016885208.1 | 16 - 17 | 1408 | 151 | |
GenBank mRNA | AB102662.1 | - | 16 - 17 | 1406 | 151 |
AB208933.1 | - | 16 - 17 | 1646 | 151 | |
AK294373.1 | - | 18 - 19 | 1539 | 151 | |
AK296775.1 | - | 15 - 16 | 1380 | 151 | |
AK313932.1 | - | 16 - 17 | 1451 | 151 | |
AK316482.1 | - | 17 - 18 | 1846 | 151 | |
BC096301.1 | - | 16 - 17 | 1447 | 151 | |
BC096302.3 | - | 16 - 17 | 1447 | 151 | |
BC096303.1 | - | 16 - 17 | 1447 | 151 | |
U08316.1 | - | 16 - 17 | 1447 | 151 |