Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RPS6KA3
- Entrez Gene ID
-
6197
- Gene Name
- ribosomal protein S6 kinase A3
- Gene Aliases
-
CLS, HU-3, ISPK-1, MAPKAPK1B, MRX19, RSK, RSK2, S6K-alpha3, p90-RSK2, pp90RSK2
- Chromosome Location
-
Chr.X: 20149911 - 20267514 on Build GRCh38
View Full Product Details