The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032591.2 | NP_115980.1 | 7 - 8 | 1179 | 70 |
GenBank mRNA | AF298591.1 | - | 7 - 8 | 1044 | 70 |