Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSPP1
Entrez Gene ID
79848
Gene Name
centrosome and spindle pole associated protein 1
Gene Aliases
CSPP, JBTS21
Chromosome Location
Chr.8: 67062417 - 67196614 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024790.6 NP_079066.5 16 - 17 2146 67
XM_005251305.3 XP_005251362.2 19 - 20 2630 67
XM_006716477.2 XP_006716540.2 14 - 15 2099 67
XM_011517598.1 XP_011515900.1 20 - 21 2672 67
XM_011517599.1 XP_011515901.1 19 - 20 2648 67
XM_011517600.1 XP_011515902.1 18 - 19 2606 67
XM_011517601.1 XP_011515903.1 18 - 19 2567 67
XM_011517602.1 XP_011515904.1 17 - 18 2525 67
XM_011517603.1 XP_011515905.1 19 - 20 2355 67
XM_011517607.1 XP_011515909.1 18 - 19 2235 67
XM_011517608.1 XP_011515910.1 17 - 18 2232 67
XM_011517609.1 XP_011515911.1 16 - 17 2354 67
XM_011517611.2 XP_011515913.1 4 - 5 731 67
XM_017013847.1 XP_016869336.1 18 - 19 2531 67
XM_017013848.1 XP_016869337.1 17 - 18 2507 67
XM_017013849.1 XP_016869338.1 17 - 18 2468 67
XM_017013850.1 XP_016869339.1 16 - 17 2426 67
XM_017013851.1 XP_016869340.1 16 - 17 2190 67
XM_017013854.1 XP_016869343.1 13 - 14 2075 67
XM_017013856.1 XP_016869345.1 11 - 12 1659 67
XM_017013857.1 XP_016869346.1 12 - 13 1709 67
XM_017013859.1 XP_016869348.1 6 - 7 859 67
GenBank mRNA AK096205.1 - 17 - 18 2200 67
AK127108.1 - 19 - 20 3675 67
AM156947.1 - 16 - 17 2147 67
BX649147.1 - 16 - 17 2147 67

View Full Product Details