Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSPP1
Entrez Gene ID
79848
Gene Name
centrosome and spindle pole associated protein 1
Gene Aliases
CSPP, JBTS21
Chromosome Location
Chr.8: 67062417 - 67196614 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291339.1 NP_001278268.1 14 - 15 2251 99
NM_024790.6 NP_079066.5 17 - 18 2251 99
XM_005251305.3 XP_005251362.2 20 - 21 2735 99
XM_006716474.2 XP_006716537.2 19 - 20 2582 99
XM_006716477.2 XP_006716540.2 15 - 16 2204 99
XM_011517598.1 XP_011515900.1 21 - 22 2777 99
XM_011517599.1 XP_011515901.1 20 - 21 2753 99
XM_011517600.1 XP_011515902.1 19 - 20 2711 99
XM_011517601.1 XP_011515903.1 19 - 20 2672 99
XM_011517602.1 XP_011515904.1 18 - 19 2630 99
XM_011517603.1 XP_011515905.1 20 - 21 2460 99
XM_011517607.1 XP_011515909.1 19 - 20 2340 99
XM_011517608.1 XP_011515910.1 18 - 19 2337 99
XM_011517609.1 XP_011515911.1 17 - 18 2459 99
XM_011517611.2 XP_011515913.1 5 - 6 836 99
XM_017013847.1 XP_016869336.1 19 - 20 2636 99
XM_017013848.1 XP_016869337.1 18 - 19 2612 99
XM_017013849.1 XP_016869338.1 18 - 19 2573 99
XM_017013850.1 XP_016869339.1 17 - 18 2531 99
XM_017013851.1 XP_016869340.1 17 - 18 2295 99
XM_017013852.1 XP_016869341.1 16 - 17 2378 99
XM_017013853.1 XP_016869342.1 16 - 17 2142 99
XM_017013854.1 XP_016869343.1 14 - 15 2180 99
XM_017013855.1 XP_016869344.1 12 - 13 1946 99
XM_017013856.1 XP_016869345.1 12 - 13 1764 99
XM_017013857.1 XP_016869346.1 13 - 14 1814 99
XM_017013858.1 XP_016869347.1 11 - 12 1562 99
XM_017013859.1 XP_016869348.1 7 - 8 964 99
GenBank mRNA AJ583433.1 - 14 - 15 2251 99
AK026143.1 - - 1244 99
AK096205.1 - 18 - 19 2305 99
AK127108.1 - 20 - 21 3780 99
AM156947.1 - 17 - 18 2252 99
BX649147.1 - 17 - 18 2252 99

View Full Product Details