The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199207.1 | NP_001186136.1 | 1 - 2 | 383 | 104 |
NM_005685.3 | NP_005676.3 | 1 - 2 | 383 | 104 | |
NM_016328.2 | NP_057412.1 | 1 - 2 | 383 | 104 | |
XM_006716182.3 | XP_006716245.1 | 1 - 2 | 179 | 104 | |
XM_011516713.1 | XP_011515015.1 | 1 - 2 | 179 | 104 | |
XM_017012802.1 | XP_016868291.1 | 1 - 2 | 179 | 104 | |
XM_017012803.1 | XP_016868292.1 | 1 - 2 | 179 | 104 | |
XM_017012805.1 | XP_016868294.1 | 1 - 2 | 179 | 104 | |
GenBank mRNA | AF151354.1 | - | 1 - 2 | 383 | 104 |
AY648295.1 | - | 1 - 2 | 203 | 104 | |
BC018136.1 | - | 1 - 2 | 148 | 104 |