The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015140.3 | NP_055955.1 | 12 - 13 | 1707 | 83 |
GenBank mRNA | AK123789.1 | - | 1 - 2 | 776 | 83 |
AK304464.1 | - | 2 - 3 | 193 | 83 | |
BC001070.2 | - | 12 - 13 | 1671 | 83 | |
CT841516.1 | - | 12 - 13 | 1648 | 83 | |
D63487.1 | - | 12 - 13 | 1629 | 83 |