The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004311.3 | NP_001004311.2 | 1 - 2 | 229 | 101 |
GenBank mRNA | AY541030.1 | - | 1 - 2 | 229 | 101 |