The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134421.2 | NP_602293.1 | 1 - 2 | 243 | 57 |
GenBank mRNA | AK000596.1 | - | 1 - 2 | 249 | 57 |