Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAPK8IP1
Entrez Gene ID
9479
Gene Name
mitogen-activated protein kinase 8 interacting protein 1
Gene Aliases
IB1, JIP-1, JIP1, PRKM8IP
Chromosome Location
Chr.11: 45885496 - 45906465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005456.3 NP_005447.1 10 - 11 2288 159
XM_005253226.3 XP_005253283.1 10 - 11 2333 159
GenBank mRNA AB209720.1 - 6 - 7 780 159
AF007134.1 - 6 - 7 564 159
AF074091.1 - 10 - 11 1963 159
AK295942.1 - 7 - 8 1409 159
BC068470.1 - 10 - 11 2133 159

View Full Product Details