Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAPK8IP1
Entrez Gene ID
9479
Gene Name
mitogen-activated protein kinase 8 interacting protein 1
Gene Aliases
IB1, JIP-1, JIP1, PRKM8IP
Chromosome Location
Chr.11: 45885496 - 45906465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005456.3 NP_005447.1 8 - 9 2105 72
XM_005253226.3 XP_005253283.1 8 - 9 2150 72
GenBank mRNA AB209720.1 - 4 - 5 597 72
AF007134.1 - 4 - 5 381 72
AF074091.1 - 8 - 9 1780 72
AK293637.1 - 7 - 8 1048 72
AK295942.1 - 5 - 6 1226 72
BC068470.1 - 8 - 9 1950 72

View Full Product Details