The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017582.6 | NP_060052.3 | 9 - 10 | 1114 | 83 |
GenBank mRNA | AF116721.1 | - | 2 - 3 | 90 | 83 |
AJ243666.1 | - | 3 - 4 | 181 | 83 | |
AK056388.1 | - | 5 - 6 | 414 | 83 | |
AK293986.1 | - | 9 - 10 | 737 | 83 | |
AY112698.1 | - | 9 - 10 | 1089 | 83 | |
AY948200.1 | - | 9 - 10 | 1097 | 83 | |
BC000848.1 | - | 5 - 6 | 448 | 83 | |
BC009286.2 | - | 4 - 5 | 252 | 83 | |
BC015316.1 | - | 1 - 2 | 57 | 83 | |
BC061583.1 | - | 5 - 6 | 426 | 83 | |
BC070158.1 | - | 8 - 9 | 639 | 83 | |
BC087836.1 | - | 7 - 8 | 525 | 83 | |
BC107057.2 | - | 10 - 11 | 782 | 83 | |
BC148185.2 | - | 9 - 10 | 791 | 83 |