Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YES1
Entrez Gene ID
7525
Gene Name
YES proto-oncogene 1, Src family tyrosine kinase
Gene Aliases
HsT441, P61-YES, Yes, c-yes
Chromosome Location
Chr.18: 721592 - 812326 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005433.3 NP_005424.1 8 - 9 1276 129
XM_005258139.3 XP_005258196.1 8 - 9 1238 129
XM_017025960.1 XP_016881449.1 8 - 9 1118 129
GenBank mRNA AK314063.1 - 8 - 9 1220 129
BC048960.1 - 8 - 9 1272 129
M15990.1 - 8 - 9 1262 129

View Full Product Details