Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HIF3A
Entrez Gene ID
64344
Gene Name
hypoxia inducible factor 3 alpha subunit
Gene Aliases
HIF-3A, HIF3-alpha-1, IPAS, MOP7, PASD7, bHLHe17
Chromosome Location
Chr.19: 46297046 - 46343433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022462.4 NP_071907.4 2 - 3 333 73
NM_152796.2 NP_690009.1 2 - 3 333 73
XM_005259153.3 XP_005259210.2 1 - 2 196 73
XM_017027139.1 XP_016882628.1 2 - 3 319 73
XM_017027140.1 XP_016882629.1 2 - 3 249 73
XM_017027141.1 XP_016882630.1 2 - 3 291 73
XM_017027142.1 XP_016882631.1 2 - 3 319 73
GenBank mRNA AK021421.1 - 2 - 3 283 73
AK297828.1 - 2 - 3 319 73
AK299699.1 - 1 - 2 243 73
DA559979.1 - 2 - 3 319 73

View Full Product Details