The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015255.2 | NP_056070.1 | 16 - 17 | 2214 | 136 |
XM_005248965.4 | XP_005249022.1 | 16 - 17 | 2331 | 136 | |
XM_005248966.3 | XP_005249023.1 | 15 - 16 | 1618 | 136 | |
XM_011514438.2 | XP_011512740.2 | 16 - 17 | 2414 | 136 | |
XM_011514439.1 | XP_011512741.1 | 7 - 8 | 781 | 136 | |
XM_011514440.1 | XP_011512742.1 | 7 - 8 | 781 | 136 | |
XM_011514441.2 | XP_011512743.1 | 4 - 5 | 394 | 136 | |
XM_017010594.1 | XP_016866083.1 | 16 - 17 | 2415 | 136 | |
XM_017010595.1 | XP_016866084.1 | 16 - 17 | 2415 | 136 | |
XM_017010596.1 | XP_016866085.1 | 16 - 17 | 2249 | 136 | |
XM_017010597.1 | XP_016866086.1 | 16 - 17 | 2416 | 136 | |
GenBank mRNA | AB002347.1 | - | 4 - 5 | 476 | 136 |
AK125795.1 | - | 8 - 9 | 880 | 136 | |
AK304070.1 | - | 8 - 9 | 867 | 136 | |
AK310532.1 | - | 7 - 8 | 908 | 136 | |
AY061884.1 | - | 16 - 17 | 1916 | 136 |